4-Hydroxyphenyl benzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09350-06
L09350-14
CAAAAL09350-06EA
115.34
CAD
CAAAAL09350-06
CAAAAL09350-14
4-Hydroxyphenyl benzoate 98%
4-Hydroxyphenyl benzoate
Purity: 98%
CAS No.: 2444-19-1
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 164-167°
Beilstein Reference: 2050596
MDL No.: MFCD00053304
Formula:
C₁₃H₁₀O₃ Melting Pt: 164…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053304 CAS Number: 2444-19-1 EINECS: 219-479-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...