10,12-Pentacosadiynoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09313-03
L09313-06
L09313-14
CAAAAL09313-03EA
93.22
CAD
CAAAAL09313-03
CAAAAL09313-06
CAAAAL09313-14
10,12-Pentacosadiynoic acid 98+%
10,12-Pentacosadiynoic acid
Purity: 98+%
CAS No.: 66990-32-7
Molecular Formula: CH3(CH2)11C≡CC≡C(CH2)8CO2H
Formula Weight: 374.61
Melting Point: 62-65°
Beilstein Reference: 4189973
Formula:
C₂₅H₄₂O₂ Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041684 CAS Number: 66990-32-7 EINECS: 614-006-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...