4-Nitrophenylhydrazine hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(4-Nitrophenyl)hydrazine hydrochloride, (4-Nitrophenyl)hydrazine hydrochloride, 4-Nitrophenylhydrazinium chloride

L09273-04 L09273-09
CAAAAL09273-04EA 51.82 CAD
CAAAAL09273-04 CAAAAL09273-09
4-Nitrophenylhydrazine hydrochloride 98%
4-Nitrophenylhydrazine hydrochloride

4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.


Purity: 98% Molecular Formula: C6H7N3O2.xHCl Formula Weight: 153.14(f.b.) Melting Point: ca 200° dec. UN No.: 2811 MDL No.: MFCD00040592

Formula: C₆H₇N₃O₂·HCl
MW: 189.6 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040592
CAS Number: 636-99-7
EINECS: 211-273-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR