N(α)-Boc-D-tryptophan 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-D-Trp-OH
L09214-03
L09214-06
CAAAAL09214-03EA
34.13
CAD
CAAAAL09214-03
CAAAAL09214-06
N(α)-Boc-D-tryptophan 97%
N(α)-Boc-D-tryptophan
Purity: 97%
CAS No.: 5241-64-5
Molecular Formula: C16H20N2O4
Formula Weight: 304.35
Storage Temperature: 2°C to 8°C
Melting Point: ca 138° dec.
Beilstein Reference: 4237334
Optical Rotation: +21° (c=1 in acetic acid)
MDL No.: MFCD00037944
Formula:
C₁₆H₂₀N₂O₄ Melting Pt: ca 138 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00037944 CAS Number: 5241-64-5 EINECS: 226-042-1 |
Specification Test Results
Optical rotation | +21° (c=1 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...