Diethyl isobutylmalonate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Isobutylmalonic Acid Diethyl Ester

L09202-06 L09202-14
CAAAAL09202-06EA 57.2 CAD
CAAAAL09202-06 CAAAAL09202-14 CAAAAL09202-22
Diethyl isobutylmalonate 98%
Diethyl isobutylmalonate

Purity: 98% CAS No.: 10203-58-4 Molecular Formula: C11H20O4 Formula Weight: 216.28 Flash Point: 100°(212°F) Refractive Index: 1.4220 Boiling Point: 225° Beilstein Reference: 1103030 MDL No.: MFCD00026869

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 225 °C
Density: 0.970
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026869
CAS Number: 10203-58-4
EINECS: 233-504-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR