Histamine dihydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09198-06
L09198-14
CAAAAL09198-06EA
87.06
CAD
CAAAAL09198-06
CAAAAL09198-14
Histamine dihydrochloride 98+%
Histamine dihydrochloride
Purity: 98+%
CAS No.: 56-92-8
Molecular Formula: C5H9N3.2HCl
Formula Weight: 184.07
Melting Point: 245-249°
Beilstein Reference: 3624116
Merck Reference: 14,4719
MDL No.: MFCD00012703
Formula:
C₅H₁₁Cl₂N₃ Melting Pt: 245…249 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012703 CAS Number: 56-92-8 EINECS: 200-298-4 Merck Index: 14,4719 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...