N-Boc-D-valine 98+%
Supplier: Thermo Scientific Chemicals
L09193-03
L09193-06
CAAAAL09193-03EA
45.19
CAD
CAAAAL09193-03
CAAAAL09193-06
N-Boc-D-valine 98+%
N-Boc-D-valine
Purity: 98+%
CAS No.: 22838-58-0
Molecular Formula: C10H19NO4
Formula Weight: 217.27
Storage Temperature: 2°C to 8°C
Melting Point: 76-80°
Beilstein Reference: 2050408
Optical Rotation: +6.3° (c=1 in acetic acid)
MDL No.: MFCD00038282
Formula:
C₁₀H₁₉NO₄ Melting Pt: 76…80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038282 CAS Number: 22838-58-0 |
Specification Test Results
Optical rotation | +6.3° (c=1 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...