3,3',5,5'-Tetramethylbiphenyl ≥97%
Supplier: Thermo Scientific Chemicals
Synonyms:
3,3',5,5'-Tetramethyl-1,1'-biphenyl
L09172-03
L09172-06
CAAAAL09172-03EA
104.91
CAD
CAAAAL09172-03
CAAAAL09172-06
3,3',5,5'-Tetramethylbiphenyl ≥97%
3,3',5,5'-Tetramethylbiphenyl
Purity: 97+%
CAS No.: 25570-02-9
Molecular Formula: C16H18
Formula Weight: 210.32
Melting Point: 47-49°
Beilstein Reference: 1935931
MDL No.: MFCD00051910
Formula:
C₁₆H₁₈ MW: 210.32 g/mol Melting Pt: 47…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051910 CAS Number: 25570-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...