Diethyl-tert-butylmalonate 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L09131-06
L09131-14
CAAAAL09131-03EA
60.81
CAD
CAAAAL09131-03
CAAAAL09131-06
CAAAAL09131-14
Diethyl-tert-butylmalonate 97%
Diethyl tert-butylmalonate
Purity: 97%
CAS No.: 759-24-0
Molecular Formula: C11H20O4
Formula Weight: 216.28
Flash Point: 90°(194°F)
Refractive Index: 1.4250
Boiling Point: 102-104°/11mm
Beilstein Reference: 1781709
MDL No.: MFCD00009152
Formula:
C₁₁H₂₀O₄ Boiling Pt: 102…104 °C/11 mm Density: 0,978 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009152 CAS Number: 759-24-0 EINECS: 212-067-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...