N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98%
Supplier: Thermo Scientific Chemicals
L09124-03
L09124-06
CAAAAL09124-03EA
34.92
CAD
CAAAAL09124-03
CAAAAL09124-06
N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98%
N-(tert-Butoxycarbonyl)-D-leucine hydrate
Purity: 98+%
CAS No.: 16937-99-8
Molecular Formula: C11H21NO4.xH2O
Formula Weight: 231.29(anhy)
Melting Point: 80-84°
Beilstein Reference: 2331060
Optical Rotation: +25° (c=2 in acetic acid)
MDL No.: MFCD00065583
Formula:
C₁₁H₂₁NO₄ Melting Pt: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038294 CAS Number: 16937-99-8 |
Specification Test Results
Optical rotation | +25° (c=2 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...