N(α)-Cbz-L-histidine 98+%

Supplier: Thermo Scientific Chemicals

L09114-03 L09114-06
CAAAAL09114-03EA 74.73 CAD
CAAAAL09114-03 CAAAAL09114-06
N(α)-Cbz-L-histidine 98+%
N(α)-Cbz-L-histidine

Purity: 98+% CAS No.: 14997-58-1 Molecular Formula: C14H15N3O4 Formula Weight: 289.29 Melting Point: ca 165° dec. Beilstein Reference: 3561682

Formula: C₁₄H₁₅N₃O₄
Melting Pt: ∼ 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00065960
CAS Number: 14997-58-1
EINECS: 239-084-0

Order Now

Specification Test Results

Optical rotation -24° (c=6 in 6N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR