DL-Homocysteinethiolactone hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl
L09077-14
L09077-22
CAAAAL09077-14EA
61.3
CAD
CAAAAL09077-14
CAAAAL09077-22
DL-Homocysteinethiolactone hydrochloride 99%
DL-Homocysteinethiolactone hydrochloride
Purity: 99%
CAS No.: 6038-19-3
Molecular Formula: C4H7NOS.HCl
Formula Weight: 153.63
Storage Temperature: 2°C to 8°C
Melting Point: ca 204° dec.
Beilstein Reference: 3562103
MDL No.: MFCD00012724
Formula:
C₄H₇NOS MW: 153.62 g/mol Melting Pt: ca 204 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012724 CAS Number: 6038-19-3 EINECS: 227-923-3 Merck Index: 14,4734 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...