Risocaine (propyl-4-aminobenzoate) 98%

Supplier: Thermo Scientific Chemicals

L09066-09 L09066-18
CAAAAL09066-09EA 63.67 CAD
CAAAAL09066-09 CAAAAL09066-18
Risocaine (propyl-4-aminobenzoate) 98%
Risocaine (Propyl 4-aminobenzoate)

Purity: 98% CAS No.: 94-12-2 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 73-75° Beilstein Reference: 1211203

Formula: C₁₀H₁₃NO₂
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00017111
CAS Number: 94-12-2
EINECS: 202-306-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR