Aminomalononitrile-p-toluenesulfonate ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Aminomalononitrile p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulfonate, Aminomalononitrile p-Toluenesulphonic acid, Aminomalononitrile p-Toluenesulfonic acid, Dicyanomethylammonium p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulphonate, Aminomalononitrile p-Toluenesulfonate, Dicyanomethylammonium p-Toluenesulfonate
Purity: 97+%
CAS No.: 5098-14-6
Molecular Formula: C10H11N3O3S
Formula Weight: 253.28
Melting Point: ca 169° dec.
Beilstein Reference: 4050747
UN No.: 3439
MDL No.: MFCD00012848
Formula:
C₃H₃N₃ MW: 253.28 g/mol Melting Pt: ∼ 169 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012848 CAS Number: 5098-14-6 EINECS: 225-817-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...