N-Carbobenzoxy-6-aminohexanoic acid 99%
Supplier: Thermo Scientific Chemicals
L08997-06
L08997-14
CAAAAL08997-06EA
86.11
CAD
CAAAAL08997-06
CAAAAL08997-14
N-Carbobenzoxy-6-aminohexanoic acid 99%
N-Carbobenzoxy-6-aminohexanoic acid
Purity: 99%
CAS No.: 1947-00-8
Molecular Formula: C6H5CH2OCONH(CH2)5CO2H
Formula Weight: 265.31
Storage Temperature: 2°C to 8°C
Melting Point: 53-57°
Beilstein Reference: 2288508
MDL No.: MFCD00004423
Formula:
C₁₄H₁₉NO₄ Melting Pt: 53…57 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004423 CAS Number: 1947-00-8 EINECS: 217-748-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...