N-(tert-Butoxycarbonyl)-D-serine ≥98%
Supplier: Thermo Scientific Chemicals
L08904-03
L08904-06
CAAAAL08904-03EA
46.45
CAD
CAAAAL08904-03
CAAAAL08904-06
N-(tert-Butoxycarbonyl)-D-serine ≥98%
N-(tert-Butoxycarbonyl)-D-serine
Purity: 98+%
CAS No.: 6368-20-3
Molecular Formula: C8H15NO5
Formula Weight: 205.21
Storage Temperature: 2°C to 8°C
Melting Point: 88-91°
Beilstein Reference: 1874714
Optical Rotation: +3.5° (c=2 in acetic acid)
MDL No.: MFCD00063142
Formula:
C₈H₁₅NO₅ Melting Pt: 88…91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063142 CAS Number: 6368-20-3 EINECS: 613-313-8 |
Specification Test Results
Optical rotation | +3.5° (c=2 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...