3-Ethylbenzothiazolium bromide 98%
Supplier: Thermo Scientific Chemicals
L08903-09
L08903-18
CAAAAL08903-09EA
108.39
CAD
CAAAAL08903-09
CAAAAL08903-18
3-Ethylbenzothiazolium bromide 98%
3-Ethylbenzothiazolium bromide
Purity: 98%
CAS No.: 32446-47-2
Molecular Formula: C9H10BrNS
Formula Weight: 244.16
Melting Point: 205-207°
Beilstein Reference: 6250086
Formula:
C₉H₁₀BrNS Melting Pt: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051852 CAS Number: 32446-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...