1,2-Di(p-tolyl)ethane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08876-03
L08876-06
CAAAAL08876-03EA
58.78
CAD
CAAAAL08876-03
CAAAAL08876-06
1,2-Di(p-tolyl)ethane 98%
1,2-Di(p-tolyl)ethane
Purity: 98%
CAS No.: 538-39-6
Molecular Formula: C16H18
Formula Weight: 210.32
Melting Point: 80-82°
Boiling Point: 296-298°/730mm
Beilstein Reference: 1909630
MDL No.: MFCD00026025
Formula:
C₁₆H₁₈ Boiling Pt: 296…298 °C/730 mm Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026025 CAS Number: 538-39-6 Merck Index: 14,3380 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...