N-Carbobenzoxy-D-valine ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cbz-D-Val-OH, Z-D-Val-OH, N-Cbz-D-valine
L08856-03
L08856-06
CAAAAL08856-03EA
146.47
CAD
CAAAAL08856-03
CAAAAL08856-06
N-Carbobenzoxy-D-valine ≥98%
N-Carbobenzoxy-D-valine
Purity: 98+%
CAS No.: 1685-33-2
Molecular Formula: C13H17NO4
Formula Weight: 251.28
Storage Temperature: 2°C to 8°C
Melting Point: 58-60°
Beilstein Reference: 2056616
Optical Rotation: +3.8° (c=2 in acetic acid)
MDL No.: MFCD00065703
Formula:
C₁₃H₁₇NO₄ MW: 251.28 g/mol Melting Pt: 58…60 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065703 CAS Number: 1685-33-2 |
Specification Test Results
Optical rotation | +3.8° (c=2 in acetic acid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...