2,2-Di-n-propylacetamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Propylpentanamide, Valpromide
L08847-06
L08847-14
CAAAAL08847-06EA
154.52
CAD
CAAAAL08847-06
CAAAAL08847-14
2,2-Di-n-propylacetamide 97%
2,2-Di-n-propylacetamide
Purity: 97%
CAS No.: 2430-27-5
Molecular Formula: C8H17NO
Formula Weight: 143.23
Odor: Odorless
Melting Point: 123-127°
Beilstein Reference: 1750444
MDL No.: MFCD00051534
Formula:
C₈H₁₇NO MW: 143.23 g/mol Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051534 CAS Number: 2430-27-5 EINECS: 219-394-2 Merck Index: 14,9914 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...