Dibenzyl phosphate 98%
Supplier: Thermo Scientific Chemicals
L08813-03
L08813-06
L08813-14
CAAAAL08813-03EA
50.24
CAD
CAAAAL08813-03
CAAAAL08813-06
CAAAAL08813-14
Dibenzyl phosphate 98%
Dibenzyl phosphate
Purity: 98%
CAS No.: 1623-08-1
Molecular Formula: C14H15O4P
Formula Weight: 278.24
Storage Temperature: 2°C to 8°C
Melting Point: 78-81°
Beilstein Reference: 2055755
MDL No.: MFCD00004775
Formula:
C₁₄H₁₅O₄P Melting Pt: 78…81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004775 CAS Number: 1623-08-1 EINECS: 216-602-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...