N(α)-Boc-D-histidine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-D-His-OH
L08810-03
L08810-MD
CAAAAL08810-03EA
114.71
CAD
CAAAAL08810-03
CAAAAL08810-MD
N(α)-Boc-D-histidine 98+%
N(α)-Boc-D-histidine
Purity: 98+%
CAS No.: 50654-94-9
Molecular Formula: C11H17N3O4
Formula Weight: 255.27
Storage Temperature: 2°C to 8°C
Melting Point: ca 197° dec.
Beilstein Reference: 4196575
Optical Rotation: -26° (c=1 in methanol)
MDL No.: MFCD00037851
Formula:
C₁₁H₁₇N₃O₄ MW: 255.3 g/mol Melting Pt: ∼ 197 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00037851 CAS Number: 50654-94-9 |
Specification Test Results
Optical rotation | -26° (c=1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...