4-(Dimethylamino)-4-oxobutanoic acid 98%
Supplier: Thermo Scientific Chemicals
L08796-06
L08796-14
CAAAAL08796-06EA
115.34
CAD
CAAAAL08796-06
CAAAAL08796-14
4-(Dimethylamino)-4-oxobutanoic acid 98%
4-(Dimethylamino)-4-oxobutanoic acid
Purity: 98%
CAS No.: 2564-95-6
Molecular Formula: C6H11NO3
Formula Weight: 145.16
Melting Point: 82-87°
Beilstein Reference: 1762788
MDL No.: MFCD00043505
Formula:
C₆H₁₁NO₃ Melting Pt: 82…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043505 CAS Number: 2564-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...