Bis(2-oxo-3-oxazolidinyl)phosphinic chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08775-06
L08775-14
CAAAAL08775-03EA
48.02
CAD
CAAAAL08775-03
CAAAAL08775-06
CAAAAL08775-14
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride 97%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
Fieser: 10,41 13,39
Purity: 97%
CAS No.: 68641-49-6
Molecular Formula: C6H8ClN2O5P
Formula Weight: 254.57
Melting Point: ca 195° dec.
Beilstein Reference: 3654596
UN No.: 3261
MDL No.: MFCD00010077
Formula:
C₆H₈ClN₂O₅P Melting Pt: ca 195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010077 CAS Number: 68641-49-6 UN: 1759 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...