5,5-Dimethyl-1,3-dioxan-2-one, tech.
Supplier: Thermo Scientific Chemicals
L08767-03
L08767-06
CAAAAL08767-03EA
58.3
CAD
CAAAAL08767-03
CAAAAL08767-06
5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one
CAS No.: 3592-12-9
Molecular Formula: C6H10O3
Formula Weight: 130.14
Melting Point: 105-114°
Beilstein Reference: 109055
MDL No.: MFCD00014650
Formula:
C₆H₁₀O₃ Melting Pt: 105…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014650 CAS Number: 3592-12-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...