3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride
L08750-14
L08750-22
CAAAAL08750-14EA
97.64
CAD
CAAAAL08750-14
CAAAAL08750-22
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Fieser: 6,38 7,16 10,27
Purity: 98%
CAS No.: 4568-71-2
Molecular Formula: C13H16ClNOS
Formula Weight: 269.80
Melting Point: 142-145°
Beilstein Reference: 3788921
MDL No.: MFCD00011959
Formula:
C₁₃H₁₆ClNOS MW: 269.79 g/mol Melting Pt: 142…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011959 CAS Number: 4568-71-2 EINECS: 224-947-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...