N-(tert-Butoxycarbonyl)-D-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH
L08722-03
L08722-06
CAAAAL08722-03EA
43.29
CAD
CAAAAL08722-03
CAAAAL08722-06
N-(tert-Butoxycarbonyl)-D-phenylalanine 98%
N-(tert-Butoxycarbonyl)-D-phenylalanine
Purity: 98%
CAS No.: 18942-49-9
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Storage Temperature: 2°C to 8°C
Melting Point: 85-88°
Beilstein Reference: 3593396
Optical Rotation: -24.5° (c=1 in ethanol)
MDL No.: MFCD00063149
Formula:
C₁₄H₁₉NO₄ Melting Pt: 85…88 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063149 CAS Number: 18942-49-9 |
Specification Test Results
Optical rotation | -24° (c=1 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...