Ethyl indole-3-acrylate 97%

Supplier: Thermo Scientific Chemicals

L08703-03 L08703-06
CAAAAL08703-03EA 77.74 CAD
CAAAAL08703-03 CAAAAL08703-06
Ethyl indole-3-acrylate 97%
Ethyl indole-3-acrylate

Purity: 97% CAS No.: 15181-86-9 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Melting Point: 118-122° Beilstein Reference: 476317 MDL No.: MFCD00051947

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00051947
CAS Number: 15181-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR