(S)-(+)-2-Hydroxy-2-phenylpropionic acid 98+%
Supplier: Thermo Scientific Chemicals
L08692-03
L08692-MD
CAAAAL08692-03EA
379.2
CAD
CAAAAL08692-03
CAAAAL08692-MD
(S)-(+)-2-Hydroxy-2-phenylpropionic acid 98+%
(S)-(+)-2-Hydroxy-2-phenylpropionic acid
Purity: 98+%
CAS No.: 13113-71-8
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 115-117°
Beilstein Reference: 2208529
Formula:
C₉H₁₀O₃ Melting Pt: 115…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067700 CAS Number: 13113-71-8 |
Specification Test Results
Optical rotation | +49° (c=2 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...