Erucic acid tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: (Z)-Docos-13-enoic acid

L08685-18 L08685-30
CAAAAL08685-18EA 62.57 CAD
CAAAAL08685-18 CAAAAL08685-30
Erucic acid tech. 90%, Technical Grade
Erucic acid

Purity: 90% CAS No.: 112-86-7 Molecular Formula: C22H42O2 Formula Weight: 338.58 Flash Point: 110°(230°F) Melting Point: 29-33° Boiling Point: 265°/15mm Beilstein Reference: 1728049 Merck Reference: 14,3674 MDL No.: MFCD00063188

Formula: C₂₂H₄₂O₂
Boiling Pt: 265 °C/15 mm
Melting Pt: 29…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063188
CAS Number: 112-86-7
EINECS: 204-011-3
Merck Index: 14,3674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR