3,4-(Methylenedioxy)benzylideneacetone 98%
Supplier: Thermo Scientific Chemicals
L08675-09
L08675-18
CAAAAL08675-09EA
104.28
CAD
CAAAAL08675-09
CAAAAL08675-18
3,4-(Methylenedioxy)benzylideneacetone 98%
3,4-(Methylenedioxy)benzylideneacetone
Purity: 98%
CAS No.: 3160-37-0
Molecular Formula: C11H10O3
Formula Weight: 190.20
Melting Point: 108-110°
Beilstein Reference: 83983
MDL No.: MFCD00016907
Formula:
C₁₁H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00016907 CAS Number: 3160-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...