Dimethyl-5-norbornene-2,3-dicarboxylate 94%

Supplier: Thermo Scientific Chemicals
Warning

L08664-09 L08664-18
CAAAAL08664-09EA 123.24 CAD
CAAAAL08664-09 CAAAAL08664-18
Dimethyl-5-norbornene-2,3-dicarboxylate 94%
Dimethyl-5-norbornene-2,3-dicarboxylate

Purity: 94% CAS No.: 5826-73-3 Molecular Formula: C11H14O4 Formula Weight: 210.23 Boiling Point: 150-152°/20mm Beilstein Reference: 1248588 MDL No.: MFCD00154455

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 150…152 °C (20 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00154455
CAS Number: 5826-73-3
Merck Index: 14,3240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR