2,2,2-Trifluoroethyl trichloromethanesulfonate 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08635-06
L08635-14
CAAAAL08635-06EA
205.4
CAD
CAAAAL08635-06
CAAAAL08635-14
2,2,2-Trifluoroethyl trichloromethanesulfonate 94%
2,2,2-Trifluoroethyl trichloromethanesulphonate
Purity: 94%
CAS No.: 23199-56-6
Molecular Formula: Cl3CSO2OCH2CF3
Formula Weight: 281.47
Refractive Index: 1.4220
Odor: Acrid
Boiling Point: 84-86°/20mm
Beilstein Reference: 1108366
UN No.: 3265
MDL No.: MFCD00042400
Formula:
C₃H₂Cl₃F₃O₃S Boiling Pt: 84…86 °C/20 mm Melting Pt: –70 °C Density: 1.714 Storage Temperature: Ambient |
MDL Number:
MFCD00042400 CAS Number: 23199-56-6 EINECS: 245-484-6 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...