N(α)-Benzyloxycarbonyl-L-lysine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH
L08629-03
L08629-06
CAAAAL08629-03EA
51.03
CAD
CAAAAL08629-03
CAAAAL08629-06
N(α)-Benzyloxycarbonyl-L-lysine 98+%
N(α)-Benzyloxycarbonyl-L-lysine
Purity: 98+%
CAS No.: 2212-75-1
Molecular Formula: C14H20N2O4
Formula Weight: 280.32
Storage Temperature: 2°C to 8°C
Melting Point: ca 237° dec.
Beilstein Reference: 2153826
Optical Rotation: -12° (c=2 in 0.2N HCl)
MDL No.: MFCD00038204
Formula:
C₁₄H₂₀N₂O₄ MW: 280.3 g/mol Melting Pt: ∼ 237 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038204 CAS Number: 2212-75-1 EINECS: 218-662-6 |
Specification Test Results
Optical rotation | -12° (c=2 in 0.2N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...