β-Cholestanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: beta-Cholestanol, 5A-Cholestan-3B-Ol, 5-ɑ-Cholestan-3-β-ol, Dihydrocholesterol

L08624-06 L08624-14
CAAAAL08624-06EA 126.24 CAD
CAAAAL08624-06 CAAAAL08624-14
β-Cholestanol 98%
beta-Cholestanol

Purity: 98% CAS No.: 80-97-7 Molecular Formula: C27H48O Formula Weight: 388.68 Melting Point: 140-144° Beilstein Reference: 2418594 Optical Rotation: +24° (c=1 in chloroform) MDL No.: MFCD00066413

Formula: C₂₇H₄₈O
MW: 388.68 g/mol
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00066413
CAS Number: 80-97-7
EINECS: 201-315-8
Merck Index: 14,2200

Order Now

Specification Test Results

Optical rotation +24° (c=1 in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR