(S)-(+)-S-Methyl-S-phenylsulfoximine 97%
Supplier: Thermo Scientific Chemicals
L08618-03
L08618-MD
CAAAAL08618-03EA
377.62
CAD
CAAAAL08618-03
CAAAAL08618-MD
(S)-(+)-S-Methyl-S-phenylsulfoximine 97%
(S)-(+)-S-Methyl-S-phenylsulfoximine
Purity: 97%
CAS No.: 33903-50-3
Molecular Formula: C7H9NOS
Formula Weight: 155.22
Melting Point: 30-32°
Beilstein Reference: 5245633
Optical Rotation: +36° (c=1 in acetone)
MDL No.: MFCD00151461
Formula:
C₇H₉NOS Melting Pt: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151461 CAS Number: 33903-50-3 |
Specification Test Results
Optical rotation | +36° (c=1 in acetone) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...