Octyltrimethylammonium bromide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trimethyl-1-octylammonium bromide
L08610-06
L08610-14
L08610-22
CAAAAL08610-06EA
65.73
CAD
CAAAAL08610-06
CAAAAL08610-14
CAAAAL08610-22
Octyltrimethylammonium bromide 97%
Octyltrimethylammonium bromide
Purity: 97%
CAS No.: 2083-68-3
Molecular Formula: CH3(CH2)7N(CH3)3Br
Formula Weight: 252.24
Melting Point: 220-222°
Beilstein Reference: 3628448
Formula:
C₁₁H₂₆BrN MW: 252.24 g/mol Melting Pt: 220…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051873 CAS Number: 2083-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...