Hexyltrimethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

L08594-06 L08594-14
CAAAAL08594-06EA 65.73 CAD
CAAAAL08594-06 CAAAAL08594-14
Hexyltrimethylammonium bromide 98%
Hexyltrimethylammonium bromide

Purity: 98% CAS No.: 2650-53-5 Molecular Formula: CH3(CH2)5N(CH3)3Br Formula Weight: 224.18 Melting Point: 177-180° Beilstein Reference: 3913229

Formula: C₉H₂₂BrN
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00051872
CAS Number: 2650-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR