3-Chloro-N,N-dimethylaniline 95%

Supplier: Thermo Scientific Chemicals
Warning

L08563-06 L08563-14
CAAAAL08563-06EA 292.3 CAD
CAAAAL08563-06 CAAAAL08563-14
3-Chloro-N,N-dimethylaniline 95%
3-Chloro-N,N-dimethylaniline

Purity: 95% CAS No.: 6848-13-1 Molecular Formula: C8H10ClN Formula Weight: 155.63 Refractive Index: 1.5760 Boiling Point: 230-232° UN No.: 2810 MDL No.: MFCD00018456

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00018456
CAS Number: 6848-13-1
EINECS: 229-935-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR