(R)-(-)-S-Methyl-S-phenylsulfoximine 97%

Supplier: Thermo Scientific Chemicals

L08562-03 L08562-MD
CAAAAL08562-03EA 448.72 CAD
CAAAAL08562-03 CAAAAL08562-MD
(R)-(-)-S-Methyl-S-phenylsulfoximine 97%
(R)-(-)-S-Methyl-S-phenylsulfoximine

Purity: 97% CAS No.: 60933-65-5 Molecular Formula: C7H9NOS Formula Weight: 155.22 Melting Point: 29-31° Beilstein Reference: 5245635 Optical Rotation: -36° (c=1 in acetone) MDL No.: MFCD00151460

Formula: C₇H₉NOS
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00151460
CAS Number: 60933-65-5

Order Now

Specification Test Results

Optical rotation -36° (c=1 in acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR