Aleuritic acid 95%
Supplier: Thermo Scientific Chemicals
L08542-06
L08542-14
CAAAAL08542-06EA
99.38
CAD
CAAAAL08542-06
CAAAAL08542-14
Aleuritic acid 95%
Aleuritic acid
Purity: 95%
CAS No.: 533-87-9
Molecular Formula: C16H32O5
Formula Weight: 304.43
Odor: Odorless
Melting Point: 97-101°
Beilstein Reference: 1727724
MDL No.: MFCD00135596
Formula:
C₁₆H₃₂O₅ Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135596 CAS Number: 533-87-9 EINECS: 208-578-8 Merck Index: 14,231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...