2-(2-Ethoxyethoxy)ethyl bromide 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2-(2-ethoxyethoxy)ethane
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃)
L08522-09
L08522-18
CAAAAL08522-09EA
316
CAD
CAAAAL08522-09
CAAAAL08522-18
2-(2-Ethoxyethoxy)ethyl bromide 98% stabilized
2-(2-Ethoxyethoxy)ethyl bromide
Stab. with potassium carbonate
Purity: 98%
CAS No.: 54550-36-6
Molecular Formula: CH3CH2OCH2CH2OCH2CH2Br
Formula Weight: 197.08
Refractive Index: 1.4530
Boiling Point: 86-88°/12mm
Beilstein Reference: 1736603
MDL No.: MFCD00051920
Formula:
CH₃CH₂OCH₂CH₂OCH₂CH₂Br MW: 197.08 g/mol Boiling Pt: 86…88 °C/12 mm Density: 1.299 Storage Temperature: Ambient |
MDL Number:
MFCD00051920 CAS Number: 54550-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...