N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
Supplier: Thermo Scientific Chemicals
L08498-03
L08498-06
CAAAAL08498-03EA
41.4
CAD
CAAAAL08498-03
CAAAAL08498-06
N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
N-(tert-Butoxycarbonyl)-L-aspartic acid
Purity: 98%
CAS No.: 13726-67-5
Molecular Formula: C9H15NO6
Formula Weight: 233.23
Storage Temperature: 2°C to 8°C
Melting Point: 115°
Beilstein Reference: 1913973
Optical Rotation: -5.5° (c=1 in methanol)
MDL No.: MFCD00037279
Formula:
C₉H₁₅NO₆ Melting Pt: 115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037279 CAS Number: 13726-67-5 EINECS: 237-294-7 |
Specification Test Results
Optical rotation | -5.5° (c=1 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...