2,4,6-Triisopropylbenzenesulfonamide 98%
Supplier: Thermo Scientific Chemicals
L08445-06
L08445-14
CAAAAL08445-06EA
101.91
CAD
CAAAAL08445-06
CAAAAL08445-14
2,4,6-Triisopropylbenzenesulfonamide 98%
2,4,6-Triisopropylbenzenesulphonamide
Purity: 98%
CAS No.: 105536-22-9
Molecular Formula: C15H25NO2S
Formula Weight: 283.44
Melting Point: 113-116°
Beilstein Reference: 2658523
Formula:
C₁₅H₂₅NO₂S Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051975 CAS Number: 105536-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...