5-Norbornene-2-carboxylic acid predominantly endo isomer 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid

L08444-06 L08444-14
CAAAAL08444-06EA 47.4 CAD
CAAAAL08444-06 CAAAAL08444-14
5-Norbornene-2-carboxylic acid predominantly endo isomer 97%
5-Norbornene-2-carboxylic acid

Purity: 97% CAS No.: 120-74-1 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 110°(230°F) Refractive Index: 1.4940 Boiling Point: 136-138°/14mm Beilstein Reference: 637167 UN No.: 3265 MDL No.: MFCD00085356

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 136…138 °C/14 mm
Density: 1.129
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00085356
CAS Number: 120-74-1
EINECS: 204-422-8
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR