2-Chloro-5-methoxybenzimidazole 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08440-03
L08440-MD
CAAAAL08440-03EA
304.94
CAD
CAAAAL08440-03
CAAAAL08440-MD
2-Chloro-5-methoxybenzimidazole 94%
2-Chloro-5-methoxybenzimidazole
Purity: 94%
CAS No.: 15965-54-5
Molecular Formula: C8H7ClN2O
Formula Weight: 182.61
Melting Point: 165-168°
Beilstein Reference: 777455
MDL No.: MFCD00051945
Formula:
C₈H₇ClN₂O Melting Pt: 165…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051945 CAS Number: 15965-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...