Norbornane-2-carboxylic acid predominantly endo isomer 98%

Supplier: Thermo Scientific Chemicals
Danger

L08439-06 L08439-14
CAAAAL08439-06EA 180.12 CAD
CAAAAL08439-06 CAAAAL08439-14
Norbornane-2-carboxylic acid predominantly endo isomer 98%
Norbornane-2-carboxylic acid

Purity: 98% CAS No.: 824-62-4 Molecular Formula: C8H12O2 Formula Weight: 140.18 Flash Point: 100°(212°F) Refractive Index: 1.4855 Melting Point: ca 40° Boiling Point: 134-136°/16mm Beilstein Reference: 2042631 UN No.: 3261 MDL No.: MFCD00167749

Formula: C₈H₁₂O₂
Boiling Pt: 134…136 °C/16 mm
Melting Pt: ca 40 °C
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00167749
CAS Number: 824-62-4
EINECS: 212-532-2
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR