N,N'-Diacetyl-1,4-phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DAPPD, N,N'-1,4-Phenylenebisacetamide, N,N'-(1,4-Phenylene)diacetamide

L08381-06 L08381-14
CAAAAL08381-06EA 78.68 CAD
CAAAAL08381-06 CAAAAL08381-14
N,N'-Diacetyl-1,4-phenylenediamine 98%
N,N'-Diacetyl-1,4-phenylenediamine

Purity: 98% CAS No.: 140-50-1 Molecular Formula: C10H12N2O2 Formula Weight: 192.22 Melting Point: 300° Beilstein Reference: 2806271 MDL No.: MFCD00026142

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00026142
CAS Number: 140-50-1
EINECS: 205-417-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR