(Phenylsulfonyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08299-03
L08299-06
L08299-14
CAAAAL08299-03EA
69.05
CAD
CAAAAL08299-03
CAAAAL08299-06
CAAAAL08299-14
(Phenylsulfonyl)acetic acid 97%
(Phenylsulfonyl)acetic acid
Purity: 97%
CAS No.: 3959-23-7
Molecular Formula: C8H8O4S
Formula Weight: 200.22
Melting Point: 109-114°
Beilstein Reference: 2504376
MDL No.: MFCD00025042
Formula:
C₈H₈O₄S Melting Pt: 109…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025042 CAS Number: 3959-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...