5-Benzoylpentanoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L08290-03 L08290-06
CAAAAL08290-03EA 49.45 CAD
CAAAAL08290-03 CAAAAL08290-06 CAAAAL08290-14
5-Benzoylpentanoic acid 99%
5-Benzoylpentanoic acid

Purity: 99% CAS No.: 4144-62-1 Molecular Formula: C12H14O3 Formula Weight: 206.24 Melting Point: 75-79° Beilstein Reference: 1961325

Formula: C₁₂H₁₄O₃
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00014380
CAS Number: 4144-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR